siRNA Details
| Gene Symbol: Lrrc4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198250.1 | NP_937893.1 | 3 | 733 | ||
| GenBank mRNA | AK140418.1 | - | 2 | 680 | ||
| BC053486.1 | - | 2 | 597 | |||
| BC060263.1 | - | 3 | 733 | 5686281 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrc4b| Gene Symbol: Lrrc4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198250.1 | NP_937893.1 | 3 | 733 | ||
| GenBank mRNA | AK140418.1 | - | 2 | 680 | ||
| BC053486.1 | - | 2 | 597 | |||
| BC060263.1 | - | 3 | 733 | 5686281 | ||