siRNA Details
| Gene Symbol: Tarsl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172310.2 | NP_758514.2 | 11 | 1381 | ||
| GenBank mRNA | AK040927.1 | - | 11 | 1354 | ||
| AK041280.1 | - | 11 | 1381 | |||
| AK044290.1 | - | 11 | 1382 | |||
| AK079416.1 | - | 11 | 1363 | |||
| BC039225.1 | - | 11 | 1365 | 4458085 | ||

Tarsl2
Genomic Map