siRNA Details
Gene Symbol: Prss43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199471.1 | NP_955765.1 | 1, 2 | 440 | ||
GenBank mRNA | AB100999.1 | - | 1, 2 | 440 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Prss43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199471.1 | NP_955765.1 | 1, 2 | 440 | ||
GenBank mRNA | AB100999.1 | - | 1, 2 | 440 |