siRNA Details
Gene Symbol: Smok4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK029728.1 | - | 2 | 1776 | ||
BC125398.1 | - | 1 | 1334 | |||
BC132092.1 | - | 1 | 1334 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Smok4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK029728.1 | - | 2 | 1776 | ||
BC125398.1 | - | 1 | 1334 | |||
BC132092.1 | - | 1 | 1334 |