siRNA Details
| Gene Symbol: Timd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178759.4 | NP_848874.3 | 2 | 228 | ||
| GenBank mRNA | AK046560.1 | - | 2 | 227 | ||
| AK079787.1 | - | 2 | 227 | |||
| AK080042.1 | - | 2 | 227 | |||
| AK080212.1 | - | 2 | 229 | |||
| AY376716.1 | - | 2 | 227 | |||
| AY376717.1 | - | 2 | 227 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BC032879.1 | - | 737 | Chr. - - [-] on Build | ||

Timd4
Genomic Map