siRNA Details
| Gene Symbol: Gemin4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177367.3 | NP_796341.2 | 2 | 584 | ||
| GenBank mRNA | AK133821.1 | - | 2 | 676 | ||
| AK141078.1 | - | 2 | 563 | |||
| AK154061.1 | - | 2 | 565 | |||
| BC031425.1 | - | 2 | 524 | 3982927 | ||
| BC062157.1 | - | 2 | 532 | 6508223 | ||

Gemin4
Genomic Map