siRNA Details
| Gene Symbol: Slfn8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167743.1 | NP_001161215.1 | 4 | 1472 | ||
| NM_181545.4 | NP_853523.2 | 6 | 2400 | |||
| GenBank mRNA | AK150875.1 | - | 4 | 1722 | ||
| AY261798.1 | - | 6 | 2400 | |||
| AY261799.1 | - | 6 | 2363 | |||
| BC152549.1 | - | 4 | 1472 | |||

Slfn8
Genomic Map