siRNA Details
| Gene Symbol: AEBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001129.4 | NP_001120.3 | 12 | 1777 | ||
| XM_011515162.1 | XP_011513464.1 | 11 | 1740 | |||
| GenBank mRNA | AB209744.1 | - | 12 | 1731 | ||
| AF053944.1 | - | 12 | 1611 | |||
| AK127541.1 | - | 1 | 66 | |||
| BC038588.1 | - | 12 | 1777 | 5764769 | ||
| D86479.1 | - | 6 | 533 | |||

AEBP1
Genomic Map