siRNA Details
| Gene Symbol: Nynrin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040072.1 | NP_001035161.1 | 3 | 1596 | ||
| GenBank mRNA | BC057379.1 | - | 1 | 47 | ||
| BC151043.1 | - | 4 | 1618 | 9087956 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nynrin| Gene Symbol: Nynrin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040072.1 | NP_001035161.1 | 3 | 1596 | ||
| GenBank mRNA | BC057379.1 | - | 1 | 47 | ||
| BC151043.1 | - | 4 | 1618 | 9087956 | ||