siRNA Details
Gene Symbol: Klhl23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177784.4 | NP_808452.2 | 3 | 1587 | ||
GenBank mRNA | AK048518.1 | - | 3 | 1486 | ||
BC072626.1 | - | 3 | 1587 | 6827841 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Klhl23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177784.4 | NP_808452.2 | 3 | 1587 | ||
GenBank mRNA | AK048518.1 | - | 3 | 1486 | ||
BC072626.1 | - | 3 | 1587 | 6827841 |