siRNA Details
| Gene Symbol: Trp53i11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025246.1 | NP_001020417.1 | 3 | 374 | ||
| GenBank mRNA | AK142316.1 | - | 4 | 547 | ||
| BC024710.1 | - | 2 | 343 | |||
| BC048398.1 | - | 3 | 286 | 6397829 | ||
| BC059041.1 | - | 3 | 246 | |||
| BC098206.1 | - | 3 | 374 | 6812412 | ||

Trp53i11
Genomic Map