siRNA Details
| Gene Symbol: Lkaaear1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199023.3 | NP_950188.2 | 2 | 309 | ||
| GenBank mRNA | AK076877.1 | - | 2 | 310 | ||
| BC087873.1 | - | 2 | 297 | 6740628 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lkaaear1| Gene Symbol: Lkaaear1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199023.3 | NP_950188.2 | 2 | 309 | ||
| GenBank mRNA | AK076877.1 | - | 2 | 310 | ||
| BC087873.1 | - | 2 | 297 | 6740628 | ||