siRNA Details
| Gene Symbol: Gm13078 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085412.2 | NP_001078881.1 | 3 | 568 | ||
| GenBank mRNA | BY725892.1 | - | 3 | 564 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm13078| Gene Symbol: Gm13078 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085412.2 | NP_001078881.1 | 3 | 568 | ||
| GenBank mRNA | BY725892.1 | - | 3 | 564 | ||