siRNA Details
Gene Symbol: Gm13088 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126325.1 | NP_001119797.1 | 3 | 714 |
Gene Symbol: Gm13103 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177571.3 | NP_808239.2 | 3 | 722 | ||
GenBank mRNA | AK054347.1 | - | 3 | 723 | ||
BC096438.1 | - | 3 | 719 | 30481183 | ||
DV662991.1 | - | 1 | 53 |
