siRNA Details
| Gene Symbol: Gm694 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033374.3 | NP_001028546.1 | 2 | 190 | ||
| GenBank mRNA | AK145525.1 | - | 2 | 191 | ||
| BC147054.1 | - | 2 | 156 | 9056056 | ||
| BC147055.1 | - | 2 | 156 | 9056077 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm694| Gene Symbol: Gm694 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033374.3 | NP_001028546.1 | 2 | 190 | ||
| GenBank mRNA | AK145525.1 | - | 2 | 191 | ||
| BC147054.1 | - | 2 | 156 | 9056056 | ||
| BC147055.1 | - | 2 | 156 | 9056077 | ||