siRNA Details
| Gene Symbol: Cyp4a12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177406.3 | NP_803125.2 | 8, 9 | 1157 | ||
| GenBank mRNA | AK144260.1 | - | 8, 9 | 1087 | ||
| BC014721.1 | - | 8, 9 | 1103 | 4240359 | ||
| BC025936.1 | - | 8, 9 | 1097 | 5097698 | ||
| BC026582.1 | - | 8, 9 | 1103 | 4971335 | ||
| BC031141.1 | - | 8, 9 | 1078 | 4972684 | ||
| BC033924.1 | - | 8, 9 | 1087 | 5101821 | ||
| CB950710.1 | - | 1, 2 | 114 | |||
| X71479.1 | - | 133 | ||||

Cyp4a12a
Genomic Map