siRNA Details
| Gene Symbol: BC049635 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177785.4 | NP_808453.1 | 4 | 893 | ||
| GenBank mRNA | AK077002.1 | - | 4 | 894 | ||
| AK133100.1 | - | 2 | 815 | |||
| BC049635.1 | - | 4 | 885 | 6774793 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BC049635| Gene Symbol: BC049635 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177785.4 | NP_808453.1 | 4 | 893 | ||
| GenBank mRNA | AK077002.1 | - | 4 | 894 | ||
| AK133100.1 | - | 2 | 815 | |||
| BC049635.1 | - | 4 | 885 | 6774793 | ||