siRNA Details
| Gene Symbol: Slc15a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177787.4 | NP_808455.2 | 5, 6 | 1141 | ||
| GenBank mRNA | AK036413.1 | - | 4, 5 | 924 | ||
| BC026899.1 | - | 2, 3 | 219 | |||
| BC039761.1 | - | 2, 3 | 230 | |||
| BC141332.1 | - | 4, 5 | 860 | 9055944 | ||
| BC141334.1 | - | 4, 5 | 860 | 9055946 | ||

Slc15a5
Genomic Map