siRNA Details
| Gene Symbol: Wfikkn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181819.2 | NP_861540.2 | 2 | 1283 | ||
| GenBank mRNA | AY308804.1 | - | 2 | 1253 | ||
| BC113144.1 | - | 2 | 1253 | 40047070 | ||
| JQ080910.1 | - | 2 | 1253 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wfikkn2| Gene Symbol: Wfikkn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181819.2 | NP_861540.2 | 2 | 1283 | ||
| GenBank mRNA | AY308804.1 | - | 2 | 1253 | ||
| BC113144.1 | - | 2 | 1253 | 40047070 | ||
| JQ080910.1 | - | 2 | 1253 | |||