siRNA Details
| Gene Symbol: AY358078 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194347.1 | NP_919328.1 | 7 | 898 | ||
| GenBank mRNA | AY358078.1 | - | 7 | 898 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AY358078| Gene Symbol: AY358078 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194347.1 | NP_919328.1 | 7 | 898 | ||
| GenBank mRNA | AY358078.1 | - | 7 | 898 | ||