siRNA Details
Gene Symbol: Stkld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198628.2 | NP_941030.2 | 7 | 532 | ||
GenBank mRNA | BC050806.1 | - | 7 | 533 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Stkld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198628.2 | NP_941030.2 | 7 | 532 | ||
GenBank mRNA | BC050806.1 | - | 7 | 533 |