siRNA Details
| Gene Symbol: Gm13083 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126324.1 | NP_001119796.1 | 2 | 651 | ||
| Gene Symbol: Pramef6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085414.2 | NP_001078883.2 | 3 | 671 | ||

Gm13083
Genomic Map