siRNA Details
| Gene Symbol: Kctd19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301173.1 | NP_001288102.1 | 9, 10 | 1435 | ||
| NM_177791.3 | NP_808459.1 | 10, 11 | 1504 | |||
| GenBank mRNA | AK076644.1 | - | 10, 11 | 1496 | ||
| BC092530.1 | - | 9, 10 | 1435 | 6744085 | ||
| BC125431.1 | - | 10, 11 | 1496 | 40129946 | ||
| BC125433.1 | - | 10, 11 | 1496 | 40129948 | ||

Kctd19
Genomic Map