siRNA Details
Gene Symbol: TWF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242397.1 | NP_001229326.1 | 8 | 852 | ||
NM_002822.4 | NP_002813.3 | 7 | 831 | |||
NR_073472.1 | 7 | 1221 | ||||
NR_073473.1 | 8 | 886 | ||||
GenBank mRNA | AB209302.1 | - | 8 | 726 | ||
AK127868.1 | - | 7 | 1091 | |||
AK291223.1 | - | 7 | 740 | |||
AK297206.1 | - | 6 | 653 | |||
BC022344.1 | - | 8 | 798 | 4249251 | ||
BC043148.1 | - | 7 | 834 | 5297068 | ||
BC068548.1 | - | 7 | 768 | |||
BT019691.1 | - | 7 | 701 | |||
CR541736.1 | - | 7 | 701 | |||
CR541761.1 | - | 7 | 701 | |||
U02680.1 | - | 7 | 761 |

More Information
Matching Taqman Gene Expression Assay:
Panther Classification:
Gene Ontology Categories:
