siRNA Details
| Gene Symbol: Pcdh19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105245.1 | NP_001098715.1 | 4 | 4372 | ||
| NM_001105246.1 | NP_001098716.1 | 3 | 4231 | |||
| GenBank mRNA | AK122492.1 | - | 4 | 2115 | ||
| BC118529.1 | - | 4 | 2420 | 40098492 | ||

Pcdh19
Genomic Map