siRNA Details
| Gene Symbol: Sbsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083903.1 | NP_001077372.1 | 1 | 261 | ||
| NM_172205.3 | NP_757342.2 | 1 | 261 | |||
| GenBank mRNA | AK081760.1 | - | 1 | 196 | ||
| BC051531.1 | - | 1 | 270 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sbsn| Gene Symbol: Sbsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083903.1 | NP_001077372.1 | 1 | 261 | ||
| NM_172205.3 | NP_757342.2 | 1 | 261 | |||
| GenBank mRNA | AK081760.1 | - | 1 | 196 | ||
| BC051531.1 | - | 1 | 270 | |||