siRNA Details
| Gene Symbol: Sbsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083903.1 | NP_001077372.1 | 1 | 393 | ||
| NM_172205.3 | NP_757342.2 | 1 | 393 | |||
| GenBank mRNA | AK003459.1 | - | 1 | 91 | ||
| AK081760.1 | - | 1 | 328 | |||
| AY115494.1 | - | 1 | 332 | |||
| AY444556.1 | - | 1 | 332 | |||
| BC051531.1 | - | 1 | 402 | |||
| BC092350.1 | - | 1 | 322 | |||
| BC139385.1 | - | 1 | 331 | 8862407 | ||

Sbsn
Genomic Map