siRNA Details
| Gene Symbol: Serpinb1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173052.2 | NP_766640.1 | 5, 6 | 682 | ||
| GenBank mRNA | AF426025.1 | - | 4, 5 | 565 | ||
| AK018226.1 | - | 5, 6 | 661 | |||
| BC030354.1 | - | 5, 6 | 627 | 5367365 | ||

Serpinb1b
Genomic Map