siRNA Details
| Gene Symbol: Serpinb1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173052.2 | NP_766640.1 | 6, 7 | 863 | ||
| GenBank mRNA | AF426025.1 | - | 5, 6 | 746 | ||
| BC030354.1 | - | 6, 7 | 808 | 5367365 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Serpinb1b| Gene Symbol: Serpinb1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173052.2 | NP_766640.1 | 6, 7 | 863 | ||
| GenBank mRNA | AF426025.1 | - | 5, 6 | 746 | ||
| BC030354.1 | - | 6, 7 | 808 | 5367365 | ||