siRNA Details
| Gene Symbol: Klk15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174865.1 | NP_777354.1 | 3 | 326 | ||
| GenBank mRNA | BC106965.1 | - | 3 | 326 | 40039744 | |
| BC106966.2 | - | 3 | 323 | |||
| BC107012.1 | - | 3 | 326 | 40047687 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klk15| Gene Symbol: Klk15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174865.1 | NP_777354.1 | 3 | 326 | ||
| GenBank mRNA | BC106965.1 | - | 3 | 326 | 40039744 | |
| BC106966.2 | - | 3 | 323 | |||
| BC107012.1 | - | 3 | 326 | 40047687 | ||