siRNA Details
| Gene Symbol: Hist3h2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178218.4 | NP_835736.1 | 1 | 163 | ||
| GenBank mRNA | AK051448.1 | - | 164 | |||
| AK077568.1 | - | 1 | 164 | |||
| AK082083.1 | - | 167 | ||||
| AK087537.1 | - | 165 | ||||
| BC058119.1 | - | 136 | 6819179 | |||
| BC063781.1 | - | 130 | 6493838 | |||

Hist3h2a
Genomic Map