siRNA Details
| Gene Symbol: Hist3h2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK051448.1 | - | 1053 | |||
| AK082083.1 | - | 1056 | ||||
| AK087537.1 | - | 1054 | ||||
| BC058119.1 | - | 1025 | 6819179 | |||
| BC063781.1 | - | 1019 | 6493838 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hist3h2a| Gene Symbol: Hist3h2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK051448.1 | - | 1053 | |||
| AK082083.1 | - | 1056 | ||||
| AK087537.1 | - | 1054 | ||||
| BC058119.1 | - | 1025 | 6819179 | |||
| BC063781.1 | - | 1019 | 6493838 | |||