siRNA Details
| Gene Symbol: Hist2h2bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175666.2 | NP_783597.2 | 1 | 104 | ||
| GenBank mRNA | BC019122.1 | - | 91 | 5003093 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hist2h2bb| Gene Symbol: Hist2h2bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175666.2 | NP_783597.2 | 1 | 104 | ||
| GenBank mRNA | BC019122.1 | - | 91 | 5003093 | ||