siRNA Details
| Gene Symbol: Hist2h2be | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178214.3 | NP_835586.2 | 1 | 262 | ||
| GenBank mRNA | AK078533.1 | - | 1 | 263 | ||
| AK090189.1 | - | 1 | 265 | |||
| AK143398.1 | - | 1 | 266 | |||
| AK162494.1 | - | 1 | 266 | |||
| AK167546.1 | - | 1 | 265 | |||
| AK168767.1 | - | 1 | 262 | |||
| AK170749.1 | - | 1 | 265 | |||
| BC047137.1 | - | 1 | 242 | 6311571 | ||
| BC058562.1 | - | 1 | 264 | |||

Hist2h2be
Genomic Map