siRNA Details
| Gene Symbol: Ankef1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175667.4 | NP_783598.1 | 7 | 1921 | ||
| GenBank mRNA | AK029804.1 | - | 2057 | |||
| BC089504.1 | - | 7 | 1962 | |||
| BC120825.1 | - | 7 | 1921 | 40129748 | ||
| BC144833.1 | - | 7 | 1936 | |||

Ankef1
Genomic Map