siRNA Details
| Gene Symbol: Gpr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175668.4 | NP_783599.1 | 2 | 1346 | ||
| GenBank mRNA | AK085829.1 | - | 2 | 1348 | ||
| AY255582.1 | - | 1 | 46 | |||
| BC099398.1 | - | 1 | 1221 | 30713585 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr4| Gene Symbol: Gpr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175668.4 | NP_783599.1 | 2 | 1346 | ||
| GenBank mRNA | AK085829.1 | - | 2 | 1348 | ||
| AY255582.1 | - | 1 | 46 | |||
| BC099398.1 | - | 1 | 1221 | 30713585 | ||