siRNA Details
| Gene Symbol: Nol4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161483.1 | NP_001154955.1 | 4, 5 | 489 | ||
| NM_199024.2 | NP_950189.2 | 4, 5 | 2259 | |||
| GenBank mRNA | AK031008.1 | - | 4, 5 | 2259 | ||
| BC056377.1 | - | 4, 5 | 482 | 6850243 | ||

Nol4
Genomic Map