siRNA Details
| Gene Symbol: Trim12c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146007.1 | NP_001139479.1 | 2 | 342 | ||
| NM_175677.4 | NP_783608.3 | 2 | 223 | |||
| GenBank mRNA | AK033764.1 | - | 2 | 343 | ||
| AK050485.1 | - | 2 | 347 | |||
| AK146437.1 | - | 2 | 302 | |||
| BC038937.1 | - | 2 | 223 | 2648191 | ||
| BC062252.1 | - | 2 | 209 | |||
| BU923784.1 | - | 2 | 223 | |||

Trim12c
Genomic Map