siRNA Details
| Gene Symbol: Fchsd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175684.4 | NP_783615.2 | 2 | 49 | ||
| GenBank mRNA | AK154732.1 | - | 2 | 83 | ||
| BC052886.1 | - | 2 | 80 | |||
| BC057367.1 | - | 2 | 49 | 6827858 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fchsd1| Gene Symbol: Fchsd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175684.4 | NP_783615.2 | 2 | 49 | ||
| GenBank mRNA | AK154732.1 | - | 2 | 83 | ||
| BC052886.1 | - | 2 | 80 | |||
| BC057367.1 | - | 2 | 49 | 6827858 | ||