siRNA Details
Gene Symbol: A230046K03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033375.2 | NP_001028547.2 | 29 | 3127 | ||
GenBank mRNA | AK038582.1 | - | 29 | 3085 | ||
AK043911.1 | - | 29 | 3098 | |||
AK129269.1 | - | 24 | 2657 | |||
AK145000.1 | - | 29 | 3109 | |||
AK161247.1 | - | 6 | 575 | |||
AK166233.1 | - | 29 | 3100 | |||
BC020314.2 | - | 26 | 2762 | |||
BC026467.1 | - | 18 | 2088 | |||
BC145215.1 | - | 29 | 3322 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_235003, XM_008765293, XM_006241205, FQ222739.
Gene Ontology Categories:
