siRNA Details
| Gene Symbol: Adgrl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198702.2 | NP_941991.1 | 9 | 1543 | ||
| GenBank mRNA | AK051766.1 | - | 8 | 1221 | ||
| AK140842.1 | - | 7 | 1382 | |||
| AK147374.1 | - | 1638 | ||||
| AK155931.1 | - | 8 | 1863 | |||
| AK157740.1 | - | 8 | 1122 | |||
| BC088989.1 | - | 9 | 1543 | 5705210 | ||
| BC094668.1 | - | 7 | 1352 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_130822, XM_008769985, XM_008769968, XM_008769981, XM_008769978, and others.
Set Membership: |
|

Adgrl3
Genomic Map