siRNA Details
| Gene Symbol: Dpep2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176913.4 | NP_795887.2 | 1 | 175 | ||
| GenBank mRNA | AF488552.1 | - | 1 | 175 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dpep2| Gene Symbol: Dpep2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176913.4 | NP_795887.2 | 1 | 175 | ||
| GenBank mRNA | AF488552.1 | - | 1 | 175 | ||