siRNA Details
| Gene Symbol: Fndc3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207636.2 | NP_997519.2 | 26 | 3660 | ||
| GenBank mRNA | AK089380.1 | - | 26 | 3625 | ||
| AK129250.1 | - | 12 | 1607 | |||
| AK154790.1 | - | 26 | 3429 | |||
| BC022140.1 | - | 3 | 290 | |||
| BC047066.1 | - | 10 | 1424 | |||
| BC066769.1 | - | 135 | ||||
| BC094678.1 | - | 17 | 2299 | |||
| BC144880.1 | - | 26 | 3468 | |||
| DQ192036.1 | - | 26 | 3661 | |||

Fndc3a
Genomic Map