siRNA Details
| Gene Symbol: Fndc3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207636.2 | NP_997519.2 | 14, 15 | 1950 | ||
| GenBank mRNA | AK032733.1 | - | 14, 15 | 1922 | ||
| AK048586.1 | - | 14, 15 | 1953 | |||
| AK089380.1 | - | 14, 15 | 1915 | |||
| AK154790.1 | - | 14, 15 | 1719 | |||
| BC094678.1 | - | 5, 6 | 590 | |||
| BC144880.1 | - | 14, 15 | 1758 | |||
| DQ192036.1 | - | 14, 15 | 1951 | |||

Fndc3a
Genomic Map