siRNA Details
| Gene Symbol: Pld5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195816.1 | NP_001182745.1 | 6 | 943 | ||
| NM_176916.4 | NP_795890.2 | 6 | 1247 | |||
| GenBank mRNA | AK046294.1 | - | 6 | 1185 | ||
| AK136851.1 | - | 5 | 1061 | |||
| AK136888.1 | - | 5 | 1061 | |||
| AK144116.1 | - | 6 | 1236 | |||
| BC100428.1 | - | 6 | 686 | 30247182 | ||

Pld5
Genomic Map