siRNA Details
| Gene Symbol: Pld5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195816.1 | NP_001182745.1 | 8, 9 | 1368 | ||
| NM_176916.4 | NP_795890.2 | 8, 9 | 1672 | |||
| GenBank mRNA | AK136851.1 | - | 7, 8 | 1486 | ||
| AK136888.1 | - | 7, 8 | 1486 | |||
| AK144116.1 | - | 8, 9 | 1661 | |||
| BC100428.1 | - | 8, 9 | 1111 | 30247182 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001191674.

Pld5
Genomic Map