siRNA Details
| Gene Symbol: Zfp672 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256516.1 | NP_001243445.1 | 4 | 1724 | ||
| NM_001256517.1 | NP_001243446.1 | 4 | 1693 | |||
| NM_001256518.1 | NP_001243447.1 | 5 | 1975 | |||
| NM_001256519.1 | NP_001243448.1 | 6 | 1904 | |||
| NM_001256520.1 | NP_001243449.1 | 3 | 1595 | |||
| NM_178761.5 | NP_848876.2 | 3 | 1563 | |||
| GenBank mRNA | AK029711.1 | - | 5 | 1980 | ||
| AK035113.1 | - | 3 | 1542 | |||
| AK085166.1 | - | 4 | 1653 | |||
| BC034706.1 | - | 4 | 1671 | 3582315 | ||

Zfp672
Genomic Map