siRNA Details
Gene Symbol: Lrtm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176920.4 | NP_795894.1 | 3 | 1316 | ||
GenBank mRNA | AK044492.1 | - | 3 | 1270 | ||
AK044756.1 | - | 1 | 114 | |||
AK138618.1 | - | 3 | 1134 | |||
BC051461.1 | - | 2 | 272 | |||
BC099587.1 | - | 3 | 1114 | 30289234 | ||
BC132594.1 | - | 3 | 1020 | 40130871 | ||
BC145196.1 | - | 3 | 913 | |||
BC145990.1 | - | 3 | 1020 | 40131293 |
