siRNA Details
| Gene Symbol: Itga11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176922.5 | NP_795896.4 | 5 | 536 | ||
| GenBank mRNA | AK054303.1 | - | 5 | 532 | ||
| AY124460.1 | - | 5 | 532 | |||
| BC058716.1 | - | 5 | 530 | 6842988 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Itga11| Gene Symbol: Itga11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176922.5 | NP_795896.4 | 5 | 536 | ||
| GenBank mRNA | AK054303.1 | - | 5 | 532 | ||
| AY124460.1 | - | 5 | 532 | |||
| BC058716.1 | - | 5 | 530 | 6842988 | ||