siRNA Details
Gene Symbol: Itga11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176922.5 | NP_795896.4 | 11 | 1355 | ||
GenBank mRNA | AK028821.1 | - | 3 | 265 | ||
BC058716.1 | - | 11 | 1349 | 6842988 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Itga11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176922.5 | NP_795896.4 | 11 | 1355 | ||
GenBank mRNA | AK028821.1 | - | 3 | 265 | ||
BC058716.1 | - | 11 | 1349 | 6842988 |